missing translation for 'onlineSavingsMsg'
Learn More

Naproxen, 99.3%, MP Biomedicals™

Product Code. 11491721 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491721 5 g 5g
11401731 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491721 Supplier MP Biomedicals Supplier No. undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22204-53-1
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.263
InChI Key CMWTZPSULFXXJA-VIFPVBQESA-N
Synonym naproxen, s-naproxen, naprosyn, naproxene, +-naproxen, equiproxen, aleve, s-+-2-6-methoxy-2-naphthyl propionic acid, laraflex, naproxeno
PubChem CID 156391
ChEBI CHEBI:7476
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O

Specifications

Color White
Assay Percent Range 0.993
Quantity 5 g
Formula Weight 230.3
Physical Form Powder
Chemical Name or Material Naproxen
compliance-icons compliance-icons
Product Identifier
  • Naproxen
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Naproxen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.