Learn More
Emma Safety Footwear Napoli Safety Shoes
Product Code. 15908181
Click to view available options
Size:
39
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15908181
Brand: Emma Safety Footwear MM11109042
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Virtually impenetrable nitrile rubber soles combined with the shoe's sophisticated looks allow you to move easily between the office and warehouse.
  • S3-Safety shoe with sole plate for wet work
  • Nitrile Rubber Heat Resistant Outsole
  • Full grain leather/Nubuck
  • ESD Certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Brown
Design Napoli
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather, Nubuck
Size 42
Toe Material Steel