missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Naphtholphthalein, pure, indicator

Product Code. 10093463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10093463 25 g Glass bottle 25g
10287153 1 g Glass bottle 1g
10286340 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10093463 Supplier Thermo Scientific Chemicals Supplier No. 190120250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 596-01-0 | C28H18O4 | 418.448 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-01-0
Molecular Formula C28H18O4
Molecular Weight (g/mol) 418.448
MDL Number MFCD00036202
InChI Key HQHBAGKIEAOSNM-UHFFFAOYSA-N
PubChem CID 68993
IUPAC Name 3,3-bis(4-hydroxynaphthalen-1-yl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=CC=C2O)C3(C4=CC=CC=C4C(=O)O3)C5=CC=C(C6=CC=CC=C65)O

Specifications

Absorbance 550 min. (at lamda max)
Melting Point 240.0°C to 250.0°C
Beilstein 18, I, 384
Merck Index 15, 6474
Infrared Spectrum Authentic
Lambda Max 648 to 654nm (in 0.1N NaOH)
Formula Weight 418.45
Loss on Drying 5% max. (110°C, 1 hr.)
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 7.3 (pinkish-yellow) to pH 8.7 (greenish--blue)
Color Beige to Pink or Brown
Quantity 25 g
Physical Form Powder
Chemical Name or Material α-Naphtholphthalein
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.