missing translation for 'onlineSavingsMsg'
Learn More

α-Naphtholbenzion, MP Biomedicals™

Product Code. 11414981 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g

Product Code. 11414981

Brand: MP Biomedicals 0215578505

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 6948-88-5
Molecular Formula C27H20O3
Molecular Weight (g/mol) 392.454
InChI Key OUYLDJXFDLBCTQ-UHFFFAOYSA-N
Synonym bis 4-hydroxy-1-naphthyl phenylmethanol, 4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, bis 4-hydroxynaphthyl phenylmethan-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, bis-4-hydroxy-1 naphthyl-phenyl-methanol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol
PubChem CID 81382
IUPAC Name 4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O

Specifications

Color Red to Brown
Quantity 5 g
Formula Weight 392.5
Physical Form Powder
Chemical Name or Material α-Naphtholbenzion
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.