missing translation for 'onlineSavingsMsg'
Learn More

Naphthol Yellow S, Thermo Scientific™

Product Code. 11439896
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439896 50 g 50g
11449896 250 g 250g
11429896 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439896 Supplier Thermo Scientific Alfa Aesar Supplier No. B20872.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 846-70-8
Molecular Formula C10H4N2Na2O8S
Molecular Weight (g/mol) 358.188
MDL Number MFCD00003958
InChI Key CTIQLGJVGNGFEW-UHFFFAOYSA-L
Synonym C.I. 10316; 5,7-Dinitro-8-hydroxy-2-naphthalenesulfonic acid disodium salt
PubChem CID 2724063
ChEBI CHEBI:87219
IUPAC Name disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate
SMILES C1=CC2=C(C=C1S(=O)(=O)[O-])C(=C(C=C2[N+](=O)[O-])[N+](=O)[O-])[O-].[Na+].[Na+]

Specifications

Melting Point >300°C
Beilstein 2708476
Merck Index 14,6393
Formula Weight 358.2
Quantity 50 g
Chemical Name or Material Naphthol Yellow S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.