missing translation for 'onlineSavingsMsg'
Learn More

Naphthol Yellow S Hydrate, TRC

Product Code. 30137432
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137432 1 g 1g
30135874 5 g 5g
30004795 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137432 Supplier Toronto Research Chemicals Supplier No. TRCN3690481G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 846-70-8
Molecular Formula C10 H4 N2 O8 S . 2 Na
Molecular Weight (g/mol) 358.1919
Synonym 2-Naphthalenesulfonic acid, 8-hydroxy-5,7-dinitro-, disodium salt (8CI,9CI), Flavianic acid sodium salt (6CI), 1307 Yellow, 1422 Yellow, A.F. Yellow No.1, Acid Leather Yellow NYS, Acid Yellow 1, Acid Yellow S, Amacid Yellow S, C.I. 10316, C.I. Acid Yellow 1, C.I. Food Yellow 1, Calcocid Yellow S, Dolkwal Naphthol Yellow S, Dynacid Yellow S, Edicol Supra Yellow Y, Ext D & C Yellow 7, Ext D and C Yellow No. 7, Ext D&C Yellow No. 7-38007, Ext D&C Yellow No. 7-90126, External D and C Yellow 7, External Yellow 7, FD and C Yellow No. 1, Flavianic acid disodium salt, Hispacid Yellow NSF, Japan Yellow 403-1, Japan Yellow No. 403-1, Java Naphthol Yellow S, KCA Naphthol Yellow S, Kiton Yellow S, Maple Naphthol Yellow S, Monacid Yellow RS, Nankai Naphthol Yellow S, Naphthol Lake Yellow FY, Naphthol Yellow, Naphthol Yellow L, Naphthol Yellow OS, Naphthol Yellow PL, Naphthol Yellow RS, Naphthol Yellow S, Naphthol Yellow S 38007, Naphthol Yellow S 90126, Naphthol Yellow SEL 12, Naphthol Yellow SFQ, Naphthol Yellow SNA, Naphthol Yellow SXX, Naphthol yellow S sodium salt, Naphtocard Yellow S, Naphtol Yellow S, Paper Yellow L, Rakuto Naphthol Yellow S, Sodium flavianate, Solar Yellow NY, Solar Yellow NY conc, Solar Yellow NY, sodium salt, Sulfur Yellow S, Transparent Yellow Lake 16091, Triacid Yellow S, Yellow 403-1, Yellow Lake 71, Yellow No. 403, Yellow No. 403-1
IUPAC Name disodium;5,7-dinitro-8-oxidonaphthalene-2-sulfonate
SMILES [Na+].[Na+].[O-]c1c(cc(c2ccc(cc12)S(=O)(=O)[O-])[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H6N2O8S.2Na/c13-10-7-3-5(21(18,19)20)1-2-6(7)8(11(14)15)4-9(10)12(16)17;;/h1-4,13H,(H,18,19,20);;/q;2*+1/p-2
Formula Weight 357.9484
Chemical Name or Material Naphthol Yellow S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.