missing translation for 'onlineSavingsMsg'
Learn More

Naphthol AS-BI phosphate, 93%

Product Code. 10127750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127750 1 g Glass bottle 1g
10474705 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10127750 Supplier Thermo Scientific Acros Supplier No. 415310010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1919-91-1 | C18H15BrNO6P | 452.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1919-91-1
Molecular Formula C18H15BrNO6P
Molecular Weight (g/mol) 452.2
MDL Number MFCD00004058
InChI Key HUXIAXQSTATULQ-UHFFFAOYSA-N
Synonym naphthol as-bi phosphate, naphthol as bi phosphate, 7-bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, 2-naphthalenecarboxamide, 7-bromo-n-2-methoxyphenyl-3-phosphonooxy, naphthol as-bi phosphoric acid, naphthol as-bi phosphoric acid solution, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl oxyphosphonic acid, 6-bromo-3-2-methoxyphenyl carbamoyl naphthalen-2-yl dihydrogen phosphate, 2-naphthalenecarboxamide,7-bromo-n-2-methoxyphenyl-3-phosphonooxy-, sodium salt 1:2, naphthol as-bl phosphate
PubChem CID 74715
IUPAC Name [6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl] dihydrogen phosphate
SMILES COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OP(=O)(O)O

Specifications

CAS Min % 91.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 91% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: partly soluble
Formula Weight 452.2
Percent Purity 93%
Physical Form Crystalline Powder
Chemical Name or Material Naphthol AS-BI phosphate
Show More Show Less
compliance-icons
Product Identifier
  • Naphthol AS-BI phosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.