missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-d8, SPEX CertiPrep™

Product Code. 15286206 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15286206 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15286206 Supplier SPEX Certiprep Supplier No. S2660

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Naphthalene-d81146-65-21000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL Amber Prescored Glass Ampule
CAS 1146-65-2, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C10H8
MDL Number MFCD00001743
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key UFWIBTONFRDIAS-PGRXLJNUSA-N
Type Certified Reference Material
SMILES [2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C([2H])=C1[2H]
IUPAC Name (2H8)naphthalene
Solvent or Matrix Methanol-P and T
Molecular Weight (g/mol) 136.22
PubChem CID 92148
Quantity 1 mL
Chemical Name or Material Naphthalene-d8
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Naphthalene-d8
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H351-Suspected of causing cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Naphthalene-d8, Methanol

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.