missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthalene-d8, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Naphthalene D8 |
| CAS | 1146-65-2 |
| Molecular Formula | C10 D8 |
| Quantity | 5 g |
| Synonym | Naphthalene-1,2,3,4,5,6,7,8-d8, Naphthalene-d8 (6 CI,8 CI,9 CI), Naphthalene (C10D8), Octadeuterionaphthalene, Octadeuteronaphthalene, Perdeuterated naphthalene, Perdeuterionaphthalene, Perdeuteronaphthalene, Naphthalene D8 |
| InChI Formula | InChI=1 S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1[2 H] |
| IUPAC Name | 1,2,3,4,5,6,7,8-octadeuterionaphthalene |
| Molecular Weight (g/mol) | 136.22 |
| Formula Weight | 136.1128 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction