missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-sulfonic acid hydrate, 94%

Product Code. 11445503
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445503 50 g 50g
11455503 250 g 250g
11435503 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445503 Supplier Thermo Scientific Alfa Aesar Supplier No. A13445.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 76530-12-6 | C10H8O3S | 208.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76530-12-6
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00149271
InChI Key KVBGVZZKJNLNJU-UHFFFAOYSA-N
Synonym naphthalene-2-sulfonic acid hydrate, 2-naphthalenesulfonic acid hydrate, 2-naphthalenesulfonic acid monohydrate, 2-naphthalenesulfonicacid, hydrate 1:1, kyselina2-naftalensulfonova, acmc-20aj4s, ksc352o8d, c10h8so3.h2o, 2-naphthalene-sulfonic acid monohydrate, naphthalene-2-sulphonic acid monohydrate
PubChem CID 20499058
IUPAC Name naphthalene-2-sulfonic acid;hydrate
SMILES OS(=O)(=O)C1=CC=C2C=CC=CC2=C1

Specifications

Quantity 50 g
UN Number UN2585
Merck Index 14,6377
Formula Weight 208.23 (Anhydrous)
Percent Purity 94%
Chemical Name or Material Naphthalene-2-sulfonic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.