missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-carbonitrile, 97%

Product Code. 11342276
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342276 5 g 5g
11352276 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342276 Supplier Thermo Scientific Alfa Aesar Supplier No. L03704.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 613-46-7 | C11H7N | 153.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-46-7
Molecular Formula C11H7N
Molecular Weight (g/mol) 153.184
MDL Number MFCD00016807
InChI Key AZKDTTQQTKDXLH-UHFFFAOYSA-N
Synonym 2-naphthonitrile, 2-cyanonaphthalene, 2-naphthalenecarbonitrile, 2-cyanonaphthylene, 2-naphthalenenitrile, beta-cyanonaphthalene, beta-naphthonitrile, .beta.-naphthonitrile, naphthalene-2-carbonitrile, 2-naphthylcyanide
PubChem CID 11944
IUPAC Name naphthalene-2-carbonitrile
SMILES C1=CC=C2C=C(C=CC2=C1)C#N

Specifications

Melting Point 63°C to 67°C
Boiling Point 156°C to 158°C (12 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 1363868
Formula Weight 153.18
Percent Purity 97%
Chemical Name or Material Naphthalene-2-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.