missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2,6-dicarboxylic acid, 98+%

Product Code. 11462898
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462898 5 g 5g
11472898 25 g 25g
11482898 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462898 Supplier Thermo Scientific Alfa Aesar Supplier No. A16868.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1141-38-4 | C12H8O4 | 216.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1141-38-4
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.192
MDL Number MFCD00004105
InChI Key RXOHFPCZGPKIRD-UHFFFAOYSA-N
Synonym 2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711
PubChem CID 14357
ChEBI CHEBI:44460
IUPAC Name naphthalene-2,6-dicarboxylic acid
SMILES C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Beilstein 2051257
Formula Weight 216.19
Percent Purity ≥98%
Chemical Name or Material Naphthalene-2,6-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.