missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,3,6-trisulfonic Acid Trisodium Salt Hydrate

Product Code. 11368858
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368858 500 g 500g
11378858 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368858 Supplier Thermo Scientific Alfa Aesar Supplier No. 041715.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123409-01-8 | C10H10Na3O10S3 | 455.33 g/mol

Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate is used as an anionic chromophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123409-01-8
Molecular Formula C10H10Na3O10S3
Molecular Weight (g/mol) 455.33
MDL Number MFCD00149263
InChI Key BMCNZRJPDRPEDI-UHFFFAOYSA-N
Synonym 1,3, 6,7-naphthalenetrisulfonic acid trisodium salt hydrate
PubChem CID 123134558
IUPAC Name naphthalene-1,3,6-trisulfonic acid;sodium;hydrate
SMILES O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O

Specifications

Quantity 500 g
Solubility Information Soluble in water.
Formula Weight 434.31 (Anhydrous)
Physical Form Powder
Chemical Name or Material Naphthalene-1,3,6-Trisulfonic Acid Trisodium Salt Hydrate
compliance-icons
Product Identifier
  • Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Naphthalene-1,3,6-trisulfonic acid trisodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.