missing translation for 'onlineSavingsMsg'
Learn More

(Naphthalen-1-yl)acetic Acid (1-Naphthylacetic Acid), Mikromol™

Product Code. 17980616
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980616 100 mg 100mg
17953706 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17980616 Supplier Mikromol Supplier No. MM0075.02
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(Naphthalen-1-yl)acetic Acid (1-Naphthylacetic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Naphthyl acetic acid
CAS 86-87-3
Analyte or Component Names 1-Naphthyl acetic acid
Shipping Condition Room Temperature
Molecular Formula C12 H10 O2
Certifications/Compliance ISO 17025
Synonym Naphazoline Hydrochloride Imp. B (EP), Naphazoline Imp. B (EP), (Naphthalen-1-yl)acetic Acid, Naphazoline Nitrate Imp. B (EP), 1-Naphthylacetic Acid, Naphazoline Hydrochloride Impurity B, Naphazoline Nitrate Impurity B, Naphazoline Impurity B, Naphazoline Impurity B
InChI Formula InChI=1S/C12H10O2/c13-12(14)8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8H2,(H,13,14)
SMILES OC(=O)Cc1cccc2ccccc12
IUPAC Name 2-naphthalen-1-ylacetic acid
Molecular Weight (g/mol) 186.21
Quantity 100 mg
Formula Weight 186.0681 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.