missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphazoline hydrochloride, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Description
Naphazoline (Naphthazoline) hydrochloride is a potent α-adrenergic receptor agonist. Naphazoline hydrochloride reduces vascular hyperpermeability and promotes vasoconstriction. Naphazoline hydrochloride reduces the levels of inflammatory factors (TNF-α, IL-1β and IL-6), cytokines (IFN-γ and IL-4), IgE, GMCSF, and NGF。Naphazoline hydrochloride can be used for non-bacterial conjunctivitis research.
Specifications
Specifications
| Chemical Name or Material | Naphazoline hydrochloride |
| Physical Form | Solid |
| CAS | 550-99-2 |
| Color | White |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C14H15ClN2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Naphthazoline hydrochloride |
| Solubility Information | H2O : 50 mg/mL (202.64 mM; Need ultrasonic) ∣DMSO : ≥ 25 mg/mL (101.32 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction