missing translation for 'onlineSavingsMsg'
Learn More

Naphazoline hydrochloride, 99%

Product Code. p-171050 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
10012380 25 g Plastic bottle 25g
10174480 100 g Plastic bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10012380

Brand: Thermo Scientific Acros 179660250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 550-99-2 | C14H14N2·ClH | 246.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550-99-2
Molecular Formula C14H14N2·ClH
Molecular Weight (g/mol) 246.74
MDL Number MFCD00012554
InChI Key DJDFFEBSKJCGHC-UHFFFAOYSA-N
Synonym naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon
PubChem CID 11079
ChEBI CHEBI:7470
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 254.0°C to 260.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Merck Index 15, 6453
Solubility Information Solubility in water: 170g/L (20°C).
Formula Weight 246.74
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Naphazoline hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.