Learn More
Invitrogen™ NAPG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589610
Description
Immunogen sequence: MAAQKINEGL EHLAKAEKYL KTGFLKWKPD YDSAASEYGK AAVAFKNAKQ FEQAKDACLR EAVAHENNRA LFHAAKAYEQ AGMMLKEMQK LPEAVQLIEK ASMMYLENGT PDTAAMALER AGKLIENVDP EKAVQLYQQT ANVFENEERL RQAVELLGKA SRLLVRGRRF DEAALSIQKE KNIYKEIENY PTCYKKTIAQ VLVHLHRNDY VAAERCVRES YSIPGFNGSE DCAALEQLLE GYDQQDQDQV SDVCNSPLFK YMDNDYAKLG LSLVVPGGGI KKKSPATPQA KPDGVTATAA DEEEDEYSGG LC; Positive Samples: SW480, Mouse spleen, Mouse lung, Rat brain; Cellular Location: Membrane, Peripheral membrane protein.
NAPG is soluble NSF attachment protein gamma. The soluble NSF attachment proteins (SNAPs) enable N-ethyl-maleimide-sensitive fusion protein (NSF) to bind to target membranes. NSF and SNAPs appear to be general components of the intracellular membrane fusion apparatus, and their action at specific sites of fusion must be controlled by SNAP receptors particular to the membranes being fused. The product of this gene mediates platelet exocytosis and controls the membrane fusion events of this process.
Specifications
| NAPG | |
| Polyclonal | |
| Unconjugated | |
| NAPG | |
| 2400003O04Rik; gamma SNAP; GAMMASNAP; gamma-soluble NSF attachment protein; NAPG; N-ethylmaleimide sensitive fusion protein attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein, gamma; NSF attachment protein gamma; SNAPG; SNAP-gamma; SNARE; soluble NSF attachment protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108123, 307382, 8774 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99747, Q9CWZ7 | |
| NAPG | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-312 of human NAPG (NP_0038171). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.