missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAPG Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598819
This item is not returnable.
View return policy
Description
NAPG Polyclonal Antibody for Western Blot, IHC (P), ELISA
NAPG is soluble NSF attachment protein gamma. The soluble NSF attachment proteins (SNAPs) enable N-ethyl-maleimide-sensitive fusion protein (NSF) to bind to target membranes. NSF and SNAPs appear to be general components of the intracellular membrane fusion apparatus, and their action at specific sites of fusion must be controlled by SNAP receptors particular to the membranes being fused. The product of this gene mediates platelet exocytosis and controls the membrane fusion events of this process.
Specifications
| NAPG | |
| Polyclonal | |
| Unconjugated | |
| NAPG | |
| 2400003O04Rik; gamma SNAP; GAMMASNAP; gamma-soluble NSF attachment protein; NAPG; N-ethylmaleimide sensitive fusion protein attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein, gamma; NSF attachment protein gamma; SNAPG; SNAP-gamma; SNARE; soluble NSF attachment protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8774 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99747 | |
| NAPG | |
| Recombinant Human Gamma-soluble NSF attachment protein (1-312AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction