missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPG Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94808-0.1ml
This item is not returnable.
View return policy
Description
NAPG Polyclonal antibody specifically detects NAPG in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NAPG | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| gamma SNAP, GAMMASNAP, gamma-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein gamma, N-ethylmaleimide-sensitive factor attachment protein, gamma, SNAPG, SNAP-gamma, soluble NSF attachment protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-312 of human NAPG (NP_003817.1). MAAQKINEGLEHLAKAEKYLKTGFLKWKPDYDSAASEYGKAAVAFKNAKQFEQAKDACLREAVAHENNRALFHAAKAYEQAGMMLKEMQKLPEAVQLIEKASMMYLENGTPDTAAMALERAGKLIENVDPEKAVQLYQQTANVFENEERLRQAVELLGKASRLLVRGRRFDEAALSIQKEKNIYKEIENYPTCYKKTIAQVLVHLHRNDYVAAERCVRESYSIPGFNGSEDCAALEQLLEGYDQQDQDQVSDVCNSPLFKYMDNDYAKLGLSLVVPGGGIKKKSPATPQAKPDGVTATAADEEEDEYSGGLC | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 8774 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction