missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAPB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116807
This item is not returnable.
View return policy
Description
Positive Samples: Mouse brain, Mouse eye, Rat brain Immunogen sequence: MDNAGKEREA VQLMAEAEKR VKASHSFLRG LFGGNTRIEE ACEMYTRAAN.
Required for vesicular transport between the endoplasmic reticulum and the Golgi apparatus.
Specifications
| NAPB | |
| Polyclonal | |
| Unconjugated | |
| NAPB | |
| beta soluble NSF attachment protein; beta-soluble NSF attachment protein; beta-soluble NSF attachment protein {ECO:0000250; brain protein 14; brain protein I47; Brp14; b-SNAP; E161; I47; NAPB; napb {ECO:0000250; N-ethylmaleimide sensitive fusion protein attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta {ECO:0000250; N-ethylmaleimide-sensitive factor attachment protein, beta; NSF attachment protein beta; Snapb; SNAP-beta; SNAP-beta {ECO:0000250; SNARE; UniProtKB:P28663} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17957, 499903, 63908 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P28663, P85969, Q9H115 | |
| NAPB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-50 of human NAPB (NP_0713631). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction