missing translation for 'onlineSavingsMsg'
Learn More

Napabucasin, MedChemExpress

Product Code. 16496356
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496356 100 mg 100mg
16451389 1 g 1g
16445918 10 mg 10mg
16432067 50 mg 50mg
16421116 200 mg 200mg
16408557 5 mg 5mg
16401716 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16496356 Supplier Medchem Express Supplier No. HY13919
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Napabucasin (BBI608) is a STAT3 inhibitor which blocks stem cell activity in cancer cells.

Napabucasin (BBI608) is a STAT3 inhibitor which blocks stem cell activity in cancer cells.

Specifications

Chemical Name or Material Napabucasin
Physical Form Powder
CAS 83280-65-3
Color Green Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H8O4
For Use With (Application) Cancer-Kinase/protease
Synonym BBI608
Solubility Information DMSO : 4.44 mg/mL (18.48 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C2OC(C(C)=O)=C1)C3=C(C2=O)C=CC=C3
Molecular Weight (g/mol) 240.21
Formula Weight 240.21
Percent Purity 99.27%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.