missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1L4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36543
This item is not returnable.
View return policy
Description
NAP1L4 Polyclonal specifically detects NAP1L4 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| NAP1L4 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| hNAP2, MGC4565, NAP-2, NAP2L, NAP2NAP1L4b, nucleosome assembly protein 1-like 4, nucleosome assembly protein 1-like 4b, Nucleosome assembly protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4676 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_005960.1 | |
| NAP1L4 | |
| The immunogen recognized by the antibody maps to a region between residue 325 to 375 of human Nucleosome Assembly Protein 2 using the numbering given in entry NP_005960.1. | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction