missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1L3 Antibody (OTI3G3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72875PCP
This item is not returnable.
View return policy
Description
NAP1L3 Monoclonal antibody specifically detects NAP1L3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| NAP1L3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 4675 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3G3 | |
| Western Blot, Immunohistochemistry | |
| BNAP, MB20, MGC26312, NPL3, nucleosome assembly protein 1-like 3 | |
| Human recombinant protein fragment corresponding to amino acids 1-315 of human NAP1L3(NP_004529) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction