missing translation for 'onlineSavingsMsg'
Learn More

NAP1L3 Antibody (OTI3G3), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18431664 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431664 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431664 Supplier Novus Biologicals Supplier No. NBP272875AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NAP1L3 Monoclonal antibody specifically detects NAP1L3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP1L3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3G3
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias BNAP, MB20, MGC26312, NPL3, nucleosome assembly protein 1-like 3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-315 of human NAP1L3 (NP_004529) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4675
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.