missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAP1L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584327
This item is not returnable.
View return policy
Description
Immunogen sequence: NELLTKTYVL KSKLAYYDPH PYRGTAIEYS TGCEIDWNEG KNVTL Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 53%.
Acidic protein which may be involved in interactions with other proteins or DNA.
Specifications
| NAP1L2 | |
| Polyclonal | |
| Unconjugated | |
| NAP1L2 | |
| Bpx; brain specific gene BPX; brain-specific protein, X-linked; Nap1l2; nucleosome assembly protein 1 like 2; nucleosome assembly protein 1-like 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4674 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULW6 | |
| NAP1L2 | |
| Recombinant protein corresponding to Human NAP1L2. Recombinant protein control fragment (Product #RP-101566). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction