Learn More
Invitrogen™ NAP1L1 Recombinant Rabbit Monoclonal Antibody (4W8B6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542881
Description
Positive test controls include: HeLa, 293T, A-431, NIH/3T3, Mouse testis, Rat lung, Rat spleen. The target is usually found in the following locations: Melanosome, Nucleus. Immunogen sequence: MADIDNKEQS ELDQDLDDVE EVEEEETGEE TKLKARQLTV QMMQNPQILA ALQERLDGLV ETPTGYIESL PRVVKRRVNA LKNLQVKCAQ IEAKFYEEVH.
NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
Specifications
| NAP1L1 | |
| Recombinant Monoclonal | |
| 0.42 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P28656, P55209, Q9Z2G8 | |
| Nap1l1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NAP1L1 (P55209). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 4W8B6 | |
| Unconjugated | |
| Nap1l1 | |
| AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; NAP1L1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; nucleosome assembly protein 1-like 1; nucleosome assembly protein-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4673, 53605, 89825 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.