missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14898-1-AP-150UL
This item is not returnable.
View return policy
Description
NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.Specifications
| NAP1L1 | |
| Polyclonal | |
| Unconjugated | |
| NAP1L1 | |
| hNRP, NAP 1 related protein, NAP1, NAP1L, NAP1L1, NRP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4673, 53605, 89825 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P28656, P55209, Q9Z2G8 | |
| Nap1l1 | |
| NAP1L1 Fusion Protein Ag6690 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction