missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511747
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.Specifications
NAP1L1 | |
Polyclonal | |
Unconjugated | |
Nap1l1 | |
AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; NAP1L1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; nucleosome assembly protein 1-like 1; nucleosome assembly protein-1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
4673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P55209 | |
Nap1l1 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human NAP1L1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |