missing translation for 'onlineSavingsMsg'
Learn More

NAP1L1 Antibody (2A9), Novus Biologicals™

Product Code. 18388697 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388697 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388697 Supplier Novus Biologicals Supplier No. H00004673M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NAP1L1 Monoclonal antibody specifically detects NAP1L1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAP1L1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2A9
Conjugate Unconjugated
Dilution Western Blot 1:100 to 1:2000, ELISA, Sandwich ELISA 1:100 to 1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004528
Gene Alias hNRP, HSP22-like protein interacting protein, MGC23410, MGC8688, NAP1, NAP-1 related protein, NAP1LFLJ16112, NRPNAP-1-related protein, nucleosome assembly protein 1-like 1
Host Species Mouse
Immunogen NAP1L1 (NP_004528, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MADIDNKEQSELDQDLDDVEEVEEEETGEETKLKARQLTVQMMQNPQILAALQERLDGLVETPTGYIESLPRVVKRRVNALKNLQVKCAQIEAKFYEEVHDLERKYAVLY
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4673
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.