missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1 Rabbit anti-Human, Mouse, Clone: 3Y6T7, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | NAP1 |
|---|---|
| Clone | 3Y6T7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18357885
|
Novus Biologicals
NBP3-15942-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18376572
|
Novus Biologicals
NBP3-15942-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAP1 Monoclonal antibody specifically detects NAP1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| NAP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Immunology, Innate Immunity | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 257397 | |
| IgG | |
| Affinity purified |
| 3Y6T7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CG5330, Dmel\CG5330, dNap, dNap1, dNAP-1, MAP3K7IP3, MGC45404, mitogen-activated protein kinase kinase kinase 7 interacting protein 3, Mitogen-activated protein kinase kinase kinase 7-interacting protein 3, Nap-1, NAP1, NF-kappa-B-activating protein 1, NFkB activating protein 1, p56/dCAF-4, TAB-3, TAK1-binding protein 3, TGF-beta activated kinase 1/MAP3K7 binding protein 3, TGF-beta-activated kinase 1 and MAP3K7-binding protein 3, TGF-beta-activated kinase 1-binding protein 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NAP1 (Q8N5C8). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title