missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAP1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82062F
This item is not returnable.
View return policy
Description
NAP1 Polyclonal antibody specifically detects NAP1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NAP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| CG5330, Dmel\CG5330, dNap, dNap1, dNAP-1, MAP3K7IP3, MGC45404, mitogen-activated protein kinase kinase kinase 7 interacting protein 3, Mitogen-activated protein kinase kinase kinase 7-interacting protein 3, Nap-1, NAP1, NF-kappa-B-activating protein 1, NFkB activating protein 1, p56/dCAF-4, TAB-3, TAK1-binding protein 3, TGF-beta activated kinase 1/MAP3K7 binding protein 3, TGF-beta-activated kinase 1 and MAP3K7-binding protein 3, TGF-beta-activated kinase 1-binding protein 3 | |
| NAP1 antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of human NAP1. The immunogen is located within amino acids 40 - 90 of NAP1. Amino Acid Squence: YHSPDDNRMNRNR | |
| 0.1 mL | |
| Apoptosis, Immunology, Innate Immunity | |
| 257397 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction