missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NANS Antibody (1G6), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97938JF549
This item is not returnable.
View return policy
Description
NANS Monoclonal antibody specifically detects NANS in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| NANS | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 1G6 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| EC 2.5.1.56, EC 2.5.1.57, N-acetylneuraminate synthase, N-acetylneuraminate-9-phosphate synthase, N-acetylneuraminic acid phosphate synthase, N-acetylneuraminic acid synthasesialic acid phosphate synthase, SASsialic acid synthase | |
| Recombinant human NANS (1-359aa) purified from E. coli | |
| 0.1 mL | |
| metabolism, Signal Transduction | |
| 54187 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction