Learn More
Invitrogen™ NANP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532941
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%); Rat, Bovine, Bat, Hamster, Panda, Pig (88%); Mouse, Dog, Elephant, Turkey, Chicken, Platypus (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
NANP (N-Acetylneuraminic Acid Phosphatase) is a Protein Coding gene. Diseases associated with NANP includeParasitic Protozoa Infectious DiseaseandIntracranial Abscess. Among its related pathways areMetabolism of proteinsandSynthesis of substrates in N-glycan biosynthesis. Gene Ontology (GO) annotations related to this gene includehydrolase activityandN-acylneuraminate-9-phosphatase activity.
Specifications
| NANP | |
| Polyclonal | |
| Unconjugated | |
| NANP | |
| 1600031M04Rik; C20orf147; dJ694B14.3; haloacid dehalogenase-like hydrolase domain containing 4; haloacid dehalogenase-like hydrolase domain-containing protein 4; Hdhd4; hypothetical protein LOC553795; N-acetylneuraminic acid phosphatase; N-acylneuraminate-9-phosphatase; NANP; nanp {ECO:0000250; Neu5Ac-9-Pase; RGD1306009; UniProtKB:Q9CPT3}; zgc:111947 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8TBE9 | |
| NANP | |
| Synthetic 16 amino acid peptide from N-terminus of human NANP. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.