missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NANP Antibody (OTI4F5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72874
This item is not returnable.
View return policy
Description
NANP Monoclonal specifically detects NANP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| NANP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000, Immunohistochemistry 1:150 | |
| C20orf147, chromosome 20 open reading frame 147, dJ694B14.3, EC 3.1.3.29, haloacid dehalogenase-like hydrolase domain containing 4, Haloacid dehalogenase-like hydrolase domain-containing protein 4, HDHD4Neu5Ac-9-Pase, MGC26833, N-acetylneuraminic acid phosphatase, N-acylneuraminate-9-phosphatase | |
| Mouse | |
| 27.6 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI4F5 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| NANP | |
| Full length human recombinant protein of human NANP(NP_689880) produced in HEK293T cell. | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 140838 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction