missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NanoVue Plus Spectrophotometer with Integrated Printer

Product Code. 10792187 Shop All Cytiva Products
Change view
Click to view available options
Pathlength:
0.2 to 0.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pathlength unitSize
10792187 0.2 to 0.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10792187 Supplier Cytiva Supplier No. 28956966

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
The NanoVue Plus spectrophotometer is an easy-to-use and reliable instrument for the measurement of nucleic acid and protein samples. Samples of 0.5μL to 5μL can be pipetted directly onto the gold-colored sample plate for measurement, and then simply recovered using a pipette. If sample recovery is not required, the sample plate can be quickly and easily wiped clean. NanoVue Plus does not require a PC for its operation and does not require time-consuming third-party recalibration. The spectrophotometer comes complete with advanced software for CyDye, incorporating intuitive dropdown lists for common fluors and the full CyDye range for easy protocol selection. NanoVue Plus also includes software for Print via Computer (PVC) and even allows you the choice of using USB cable, Bluetooth wireless connectivity, integrated printer option, or SD card. Data output is via standard USB cable and PVC (print via computer ) software, allowing data to be transferred to a PC or with the integrated printer, wireless Blutooth or SD card options.
  • UV/Visible spectrophotometer for quick and accurate quantitation of nucleic acids and protein samples.
  • Convenient, drop and read sample plate eliminates the need for cuvettes and capillaries.
  • Gold-colored sample plate enhances compatibility with the wide range of chemicals and sample types used in today's busy laboratories.
  • Accurately assays low volumes of 0.5 to 5μL, conserving precious samples.
  • Hydrophobic coating reduces sample carry over and allows for excellent sample recovery.
  • Predefined methods for nucleic acid quantitation, including concentration and purity.
  • Predefined methods for protein quantitation, including direct UV, Bradford, BCA, Biuret, and Lowry, with capability to run up to 27 standards.
  • No PC: Large, high-resolution graphical display enables a quick read of relevant results.
  • Calibration curves, kinetics or ratio measurements are all displayed at the touch of a button.
TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range ±1% at 259 nm at 0.7 to 0.8 A
Type Spectrophotometer with Integrated Printer
Electrical Requirements 100/240 V ± 10%, 50/60 Hz, 50 V
Pathlength 0.2 to 0.5 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.