Learn More
Cytiva (formerly Pall Lab) Nanosep™ Centrifugal Devices with Omega™ Membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
Brand:  Cytiva (formerly Pall Lab) OD300C35
1621.77 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.
Additional Details : Weight : 1.32050kg
 View more versions of this product
Product Code. 17174791
  • £1460.00 / Pack of 500
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • Simple, reliable,concentrating and desalting of 50 to 500 μL samples
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Orange
PES
Non-sterile
500/Pk.
EFA: 0.3 cm2
300 kDa
Polypropylene
50 to 500 μL (Sample)