Learn More
Cytiva NAB Nanosep™ Centrifugal Devices for Nucleic Acid Isolation
Click to view available options
Includes:
100/Pk.
24/Pk.
Unit Size:
Pack of 100
Pack of 24
Description
Don't let left-over reagents go to waste, use them with the Nucleic Acid Binding™ universal spin device. This centrifugal device can also be used with buffers prepared inhouse or off-the-shelf. A variety of protocols are available to download for different starting materials.
Downstream Applications:
Purified Plasmid DNA
- Restriction digest
- Cloning
- Sanger sequencing
Purified Genomic DNA
- qPCR
- Next Generation Sequencing
Purified Total RNA
- RT-PCR
- cDNA library construction
This product is not approved for use in medical, clinical, surgical or other patient applications.
Specifications
Specifications
| Description | EFA: 0.3 cm2 |
| Material (Membrane) | Glass Fiber |
| Sterility | Non-sterile |
| Housing Material | Polypropylene |
| Includes | 24/Pk. |
| Volume (Metric) | 50 to 500 μL |