missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NANOS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21679-1-AP-150UL
This item is not returnable.
View return policy
Description
os is a zinc-finger containing, RNA-binding protein that has been implicated in germ cell development in both invertebrates and vertebrates. In Drosophila, os represses apoptosis during development to ensure proper germ-line development. Unlike os1 whose expression in mice is dispensable, the os2 and os3 proteins are required for germ cell development. os2-null primordial germ cells (PGCs) die only in the male gonads and show no defects in females, while os3-null PGCs are lost during the migration stage regardless of sex. os2 and os3 have distinct expression patterns during embryo development, suggesting that these two proteins do not have redundant functions. However, expression of os2 can at least partially replace os3 function in a os3-null background. os3 expression can not rescue os2-null defects.Specifications
| NANOS3 | |
| Polyclonal | |
| Unconjugated | |
| NANOS3 | |
| os homolog 3, os homolog 3 (Drosophila), NANOS1L, NANOS3, NOS 3, NOS3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 244551, 288909, 342977 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P60323, P60324 | |
| os3 | |
| NANOS3 Fusion Protein Ag16014 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction