missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nanos3 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76919PCP
This item is not returnable.
View return policy
Description
Nanos3 Polyclonal antibody specifically detects Nanos3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Nanos3 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| MGC120114, nanos homolog 3, nanos homolog 3 (Drosophila), NOS-3, NOS3NANOS1L | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of the human Nanos3. The immunogen is located within the last 50 amino acids of Nanos3. Amino Acid Squence: CKLVRPDKAKQDTGHR | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Stem Cells | |
| 342977 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction