missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NANOS1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12858732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858732 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858732 Supplier Invitrogen™ Supplier No. PA520556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-20556 can be used with blocking peptide PEP-0676.

NANOS1 may regulate translation of specific mRNAs by forming a complex with PUM2 that associates with the 3'-UTR of mRNA targets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NANOS1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NANOS1
Gene Accession No. Q8WY41
Gene Alias EC_Rep1a; nanos 1; nanos C2HC-type zinc finger 1; nanos homolog 1; nanos homolog 1 (Drosophila); Nanos1; Nos; NOS1; NOS-1; RP11-498J9.3; SPGF12; ZC2HC12A
Gene Symbols NANOS1
Host Species Rabbit
Immunogen A 17 amino acid peptide from near the amino terminus of human Nanos1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340719
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.