missing translation for 'onlineSavingsMsg'
Learn More

NANOGP8 Polyclonal Antibody, Invitrogen™

Product Code. 15556121
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15556121 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15556121 Supplier Invitrogen Supplier No. PA539211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This locus is a processed pseudogene of the transcription factor NANOG. NANOG plays a central role in regulating self-renewal in pluripotent stem cells and tumor cells. This pseudogene contains an intact open reading frame that could potentially encode a protein similar to NANOG. Although there is no evidence of transcription from this pseudogene, RT-PCR studies suggest that NANOGP8 may be expressed in some cancer cell lines. In vitro studies using a recombit NANOGP8 protein have shown that the protein localizes to the nucleus and can promote cell proliferation, similar to NANOG. [provided by RefSeq, Sep 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NANOGP8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NANOGP8
Gene Accession No. Q6NSW7
Gene Symbols NANOGP8
Host Species Rabbit
Immunogen A synthetic peptide derived from the internal region of human NANOGP8
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388112
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.