missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NanoDrop™ Reagents

Product Code. 31921103 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
PV-8 Performance Verification
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
31921103 PV-1 Performance Verification Each
31921070 CF-1 Calibration Check Fluid Each
31921052 CF-8 Calibration Check Kit Each
31921051 ND PR-1 Reconditioning Kit Each
30089480 PV-8 Performance Verification Each
31921081 PV-8 Performance Verification Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31921103 Supplier Thermo Scientific™ Supplier No. CHEMPV1
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified standards for verifying instrument performance to factory specifications are available separately.

Features include:
  • Solution with certified absorbance values
  • Use to create a record of correct photometric performance of a NanoDrop One spectrophotometer

Specifications

Type PV-1 Performance Verification
For Use With (Equipment) NanoDrop Ultra, NanoDrop One and NanoDrop OneC spectrophotometers
Storage Requirements Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.