Learn More
Thermo Scientific NanoDrop™ Reagents
Product Code. 31921070 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
PV-8 Performance Verification
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
31921070 CF-1 Calibration Check Fluid Each
31921052 CF-8 Calibration Check Kit Each
31921051 ND PR-1 Reconditioning Kit Each
31921103 PV-1 Performance Verification Each
30089480 PV-8 Performance Verification Each
31921081 PV-8 Performance Verification Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31921070 Supplier Thermo Scientific Supplier No. CHEMCF1
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Certified standards for verifying instrument performance to factory specifications are available separately.
For use with Thermo Scientific NanoDrop Spectrophotometers.
  • Ten times more concentrated compared to other K2Cr2O7 calibration solutions
  • Packaged in 0.5mL snap-top glass ampules
  • Intended for single use only
Specifications
Type CF-1 Calibration Check Fluid
For Use With (Equipment) NanoDrop 8000, NanoDrop Lite, NanoDrop 2000, NanoDrop 2000c spectrophotometers
Packaging 0.5 mL snap-top glass ampules