Learn More
Thermo Scientific NanoDrop™ Reagents
Product Code. 31921051 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
CF-1 Calibration Check Fluid
CF-8 Calibration Check Kit
ND PR-1 Reconditioning Kit
PV-1 Performance Verification
PV-8 Performance Verification
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
31921051 ND PR-1 Reconditioning Kit Each
31921070 CF-1 Calibration Check Fluid Each
31921052 CF-8 Calibration Check Kit Each
31921103 PV-1 Performance Verification Each
30089480 PV-8 Performance Verification Each
31921081 PV-8 Performance Verification Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31921051 Supplier Thermo Scientific Supplier No. CHEMPR1KIT
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Certified standards for verifying instrument performance to factory specifications are available separately.
This kit is useful for reconditioning the sample measurement pedestals to a hydrophobic state if they become “unconditioned” (see user manual for more information). The PR-1 kit consists of a small screw cap vial of specially formulated compound and a supply of 25 convenient applicators. Kit sold separately from instrument.
For use with Thermo Scientific™ NanoDrop™ Spectrophotometers.
Includes:
Small screw cap vial of specially formulated reconditioning compound and 25 convenient swabs
Spécification
Type ND PR-1 Reconditioning Kit
For Use With (Equipment) All Thermo Scientific NanoDrop Spectrophotometers
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.