Learn More
Thermo Scientific™ NanoDrop™ Eight Tube Rack
Product Code. 31921084 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop Eight Spectrophotometers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31921084 NanoDrop Eight Spectrophotometers Each
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31921084 Supplier Thermo Scientific™ Supplier No. 840343700
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
The Thermo Scientific™ NanoDrop™ Eight Tube Rack allows you to place samples in tubes on the instrument LED board to help you keep track of samples measured.
The Thermo Scientific™ NanoDrop™ Eight Tube Rack allows you to place samples in tubes on the instrument LED board to help you keep track of samples measured.
This single tube rack allows users to keep track of sample measurements for samples stored in 0.5 mL and 1.5 mL-sized tubes.
Specifications
Description NanoDrop Eight Tube Rack
For Use With (Equipment) NanoDrop Eight Spectrophotometers