Learn More
Thermo Scientific™ NanoDrop Label Printer - DYMO LabelWriter 550
Shop All Thermo Scientific Products
Click to view available options
Description:
DYMO LabelWriter 550 Turbo w/Labels
Description
The Dymo LabelWriter 550 Turbo label printer with labels for the Thermo Scientific™ NanoDrop™ is a convenient accessory for our microvolume UV-Vis instruments. It quickly and easily prints sample data onto self-adhesive labels, which are included with purchase of printer.
Features include:
- Generates 2.31 x 4 inch (59mm x 102mm) self-adhesive labels, allowing you to place labels directly in laboratory notebooks
- Prints results for single samples or batches of samples
- Direct thermal label printer, no ink or toner required
LabelWriter is a registered trademark of DYMO Corporation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| For Use With (Equipment) | NanoDrop 2000, NanoDrop 8000, NanoDrop 2000c, NanoDrop OneC, NanoDrop One |
| Description | DYMO LabelWriter 550 Turbo w/Labels |