missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nanocon™ Crossflow Concentrator, 30 nm, 40 mL

Product Code. 30485025 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6 Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30485025 6 Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30485025 Supplier Thermo Scientific™ Supplier No. N003040
  • ONLINE EXCLUSIVE PRICE
    £332.00
    £288.84 / Pack of 1
    Save £43.16
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Thermo Scientific Nanocon Crossflow Concentrator is an advanced concentration device designed for the enrichment of nano-sized particles, including exosomes, lipid nanoparticles, and viruses. The concentrator's versatility allows for processing various biological fluids, such as urine and cell culture media, making it ideal for large sample volumes across a broad range of research applications.

The Thermo Scientific Nanocon Crossflow Concentrator is an advanced concentration device designed for the enrichment of nano-sized particles, including exosomes, lipid nanoparticles, and viruses. Utilizing cutting-edge tangential flow centrifugation technology with a pre-treated 30-nm membrane featuring tight pore distribution, this device achieves superior enrichment while minimizing sample loss. The concentrator's versatility allows for processing various biological fluids, such as urine and cell culture media, making it ideal for large sample volumes across a broad range of research applications.

  • Cutting-edge technology—leverage the latest advancements in tangential flow centrifugation to stay ahead in the competitive field of exosome research
  • High-quality enrichment—specifically designed to ensure reliable and reproducible results without compromising sample integrity
  • Optimized recovery—maximize yield with efficient separation processes that minimize sample loss and enhance recovery rates
  • Broad application range—ideal for various biological fluids, including urine and cell culture media, making it a versatile tool for enrichment from large sample volumes

The Nanocon Concentrator is engineered to handle a maximum internal sample volume of 40 mL, making it ideal for concentrating samples of 40 mL or greater, and are compatible with swinging-bucket rotors. The concentrator fits perfectly into inserts that accommodate 250-mL bottles. The recommended maximum centrifugal force is 2,000 x g, providing an ideal balance between efficient concentration and sample integrity.

Simply add sample to the device and spin. Enriched sample can be prepped using EasyPep MS sample prep kits and analyzed by mass spectrometry. If desired, additional isolation can be performed using Exosome isolation reagents (Cat. Nos. 10616D, 10614D, and 10606D) for downstream analysis by western blot, mass spectrometry, flow cytometry, or transmission electron microscopy.

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Centrifuge Filter
Format Bottle, Spin Column
Material Polystyrene, Polyethylene, SAN
Pore Size 30 nm
Max. Force 2000 x g
Quantity 6 Pk.
Sample Type Cell culture supernatant, urine
Type Gamma Irradiated Concentrator
For Use With (Equipment) Centrifuge
Sample Volume ≥ 40 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.