missing translation for 'onlineSavingsMsg'
Learn More

Melford NANA

Product Code. 30017936
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017936 100 mg 100mg
30017935 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30017936 Supplier Melford Supplier No. A100800.1
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for N-Acetylneuraminic acid aldolase.

Substance Name: n-Acetylneuraminic acid, Synthetic Sialic Acid

CAS Number: 131-48-6

Molecular Formula: C11H19NO9

Molecular Weight: 309.2

Chemical Purity:>99%

Soluble in:Water

Store at:-20°C

Specifications

CAS 131-48-6
Molecular Formula C11H19NO9
Quantity 100 mg
Molecular Weight (g/mol) 309.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.