missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NAN-190 hydrobromide
GREENER_CHOICE

Product Code. 18065862
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18065862

Brand: Tocris Bioscience™ 0553/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-HT1A antagonist

5-HT1A antagonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115338-32-4
Molecular Formula C23H28BrN3O3

Specifications

Quantity 50 mg
Formula Weight 474.4
Purity >98%
Molecular Weight (g/mol) 474.399
Chemical Name or Material NAN-190 hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.