missing translation for 'onlineSavingsMsg'
Learn More

NAN-190 hydrobromide, MedChemExpress

Product Code. p-200099731
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497726 100 mg 100mg
16457928 10 mg 10mg
16453867 50 mg 50mg
16441296 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497726 Supplier Medchem Express Supplier No. HY19818A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist. NAN-190 is a selective antagonist of 5-HT1A.

NAN-190 hydrobromide is a serotonin receptor 5-HT antagonist. NAN-190 is a selective antagonist of 5-HT1A.

Specifications

Chemical Name or Material NAN-190 hydrobromide
CAS 115338-32-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H28BrN3O3
Quantity 100 mg
Solubility Information DMSO : 16.67 mg/mL (35.14 mM; Need ultrasonic)
SMILES O=C1N(CCCCN2CCN(C3=CC=CC=C3OC)CC2)C(C4=C1C=CC=C4)=O.[H]Br
Molecular Weight (g/mol) 474.39
Formula Weight 474.39
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.